missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diisopropylsalicylic Acid, TRC

Product Code. 30381302
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381302 1 g 1g
30385457 5 g -
30377579 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381302 Supplier Toronto Research Chemicals Supplier No. TRCD5254451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2215-21-6
Molecular Formula C13H18O3
Molecular Weight (g/mol) 222.28
Synonym 2-Hydroxy-3,5-bis(1-methylethyl)benzoic Acid, 2-Hydroxy-3,5-bis(propan-2-yl)benzoic Acid, 2-Hydroxy-3,5-diisopropylbenzoic Acid, 3,5-Diisopropyl-2-hydroxybenzoic Acid
IUPAC Name 2-hydroxy-3,5-di(propan-2-yl)benzoic acid
SMILES CC(C)c1cc(C(C)C)c(O)c(c1)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H18O3/c1-7(2)9-5-10(8(3)4)12(14)11(6-9)13(15)16/h5-8,14H,1-4H3,(H,15,16)
Formula Weight 222.1256
Chemical Name or Material 3,5-Diisopropylsalicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.