missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Diiodosalicylic Acid, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 3,5-Diiodosalicylic Acid |
| CAS | 133-91-5 |
| Molecular Formula | C7H4I2O3 |
| Quantity | 25 g |
| Synonym | 2-Hydroxy-3,5-diiodo-benzoic Acid, 2-Hydroxy-3,5-diiodobenzenecarboxylic acid, 2-Hydroxy-3,5-diiodobenzoic Acid, 3,5-Diiodo-2-hydroxybenzoic Acid, 3,5-Diiodosalicylic Acid, NSC 6303 |
| InChI Formula | InChI=1S/C7H4I2O3/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2,10H,(H,11,12) |
| SMILES | OC(=O)c1cc(I)cc(I)c1O |
| Molecular Weight (g/mol) | 389.91 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction