missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodosalicylic acid, 97%

Product Code. 11448657
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11448657

Brand: Thermo Scientific Alfa Aesar A19786.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 133-91-5 | C7H4I2O3 | 389.915 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-91-5
Molecular Formula C7H4I2O3
Molecular Weight (g/mol) 389.915
MDL Number MFCD00002444
InChI Key DHZVWQPHNWDCFS-UHFFFAOYSA-N
Synonym 3,5-diiodosalicylic acid, 3,5-diiodo-2-hydroxybenzoic acid, benzoic acid, 2-hydroxy-3,5-diiodo, 2-hydroxy-3,5-diiodobenzoate, 3,5-diiodosalicylate, 2-hydroxy-3,5-diiodobenzenecarboxylic acid, salicylic acid, 3,5-diiodo, 2-hydroxy-3,5-diiodo-benzoic acid, benzoic acid, 3,5-diiodo-2-hydroxy, 3,5-diiodosalicylicacid
PubChem CID 8631
IUPAC Name 2-hydroxy-3,5-diiodobenzoic acid
SMILES C1=C(C=C(C(=C1I)O)C(=O)O)I

Specifications

Melting Point 228°C to 234°C
Quantity 500 g
Beilstein 2615358
Sensitivity Light sensitive
Merck Index 14,3190
Formula Weight 389.91
Percent Purity 97%
Chemical Name or Material 3,5-Diiodosalicylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.