missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodosalicylaldehyde, 98+%

Product Code. 11347595
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347595 50 g 50g
11337595 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347595 Supplier Thermo Scientific Alfa Aesar Supplier No. L05783.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2631-77-8 | C7H4I2O2 | 373.916 g/mol

3,5-Diiodosalicylaldehyde is used to synthesize new Schiff bases, (2,4-diiodo-6-[(2-morpholin-4-yl-ethylimino)-methyl]-phenol and 2,4-diiodo-6-[(3-morpholin-4-yl-propylimino)-methyl]-phenol) and the new tridentate ligand, [(2-hydroxy-3,5-diiodo-benzylidene)-amino]-acetic acid (HDBA), 3-bromo-N?-(2-hydroxy-3,5-diiodobenzylidene)benzohydrazide monohydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Diiodosalicylaldehyde is used to synthesize new Schiff bases, (2,4-diiodo-6-[(2-morpholin-4-yl-ethylimino)-methyl]-phenol and 2,4-diiodo-6-[(3-morpholin-4-yl-propylimino)-methyl]-phenol) and the new tridentate ligand, [(2-hydroxy-3,5-diiodo-benzylidene)-amino]-acetic acid (HDBA), 3-bromo-N′-(2-hydroxy-3,5-diiodobenzylidene)benzohydrazide monohydrate.

Solubility
Insoluble in water.

Notes
Air & Light Sensitive. Store in the dark. Incompatible with light and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2631-77-8
Molecular Formula C7H4I2O2
Molecular Weight (g/mol) 373.916
MDL Number MFCD00003321
InChI Key MYWSBJKVOUZCIA-UHFFFAOYSA-N
PubChem CID 75829
IUPAC Name 2-hydroxy-3,5-diiodobenzaldehyde
SMILES C1=C(C=C(C(=C1I)O)C=O)I

Specifications

Melting Point 106°C to 110°C
Quantity 50 g
Beilstein 1102552
Sensitivity Air and light sensitive
Synonym 3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo
Solubility Information Insoluble in water.
Formula Weight 373.92
Percent Purity ≥98%
Chemical Name or Material 3,5-Diiodosalicylaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.