missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-L-tyrosine hydrate, 98% (dry wt.), may contain up to ca 10% water, Thermo Scientific Chemicals

Product Code. 11423474
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423474 5 g 5g
11433474 25 g 25g
11443474 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423474 Supplier Thermo Scientific Alfa Aesar Supplier No. A10702.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18835-59-1
Molecular Formula C9H13I2NO5
Molecular Weight (g/mol) 469.01
MDL Number MFCD00150275
InChI Key YWAGQOOMOOUEGY-QVGJJETENA-N
Synonym 3,5-diiodo-l-tyrosine dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, 3,5-diiodotyrosine dihydrate, 3,5-dijodtyrosindihydrat, c9h9i2no3.2h2o, 3,5-diiodo-l-tyrosine hydrate, l-tyrosine,3,5-diiodo-, hydrate 1:2, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoicaciddihydrate, 2s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl-propanoic acid dihydrate
PubChem CID 2724330
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid;dihydrate
SMILES O.O.N[C@@H](CC1=CC(I)=C(O)C(I)=C1)C(O)=O

Specifications

Melting Point 185°C (decomposition)
Quantity 5 g
Formula Weight 432.98 (Anhydrous)
Sensitivity Light sensitive
Percent Purity 98%
Concentration or Composition (by Analyte or Components) May contain up to ≈10% water
Assay (dry wt.)
Chemical Name or Material 3,5-Diiodo-L-tyrosine hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.