Learn More
3,5-Diiodo-L-tyrosine hydrate, 98% (dry wt.), may contain up to ca 10% water, Thermo Scientific Chemicals
£22.30 - £176.00
Chemical Identifiers
CAS | 18835-59-1 |
---|---|
Molecular Formula | C9H13I2NO5 |
Molecular Weight (g/mol) | 469.01 |
MDL Number | MFCD00150275 |
InChI Key | YWAGQOOMOOUEGY-QVGJJETENA-N |
Synonym | 3,5-diiodo-l-tyrosine dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, 3,5-diiodotyrosine dihydrate, 3,5-dijodtyrosindihydrat, c9h9i2no3.2h2o, 3,5-diiodo-l-tyrosine hydrate, l-tyrosine,3,5-diiodo-, hydrate 1:2, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoicaciddihydrate, 2s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl-propanoic acid dihydrate |
PubChem CID | 2724330 |
IUPAC Name | (2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid;dihydrate |
SMILES | O.O.N[C@@H](CC1=CC(I)=C(O)C(I)=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11423474
|
Thermo Scientific Alfa Aesar
A10702.06 |
5 g |
£22.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433474
|
Thermo Scientific Alfa Aesar
A10702.14 |
25 g |
£58.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443474
|
Thermo Scientific Alfa Aesar
A10702.22 |
100 g |
£176.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18835-59-1 | |
469.01 | |
YWAGQOOMOOUEGY-QVGJJETENA-N | |
2724330 | |
O.O.N[C@@H](CC1=CC(I)=C(O)C(I)=C1)C(O)=O |
C9H13I2NO5 | |
MFCD00150275 | |
3,5-diiodo-l-tyrosine dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, 3,5-diiodotyrosine dihydrate, 3,5-dijodtyrosindihydrat, c9h9i2no3.2h2o, 3,5-diiodo-l-tyrosine hydrate, l-tyrosine,3,5-diiodo-, hydrate 1:2, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoicaciddihydrate, 2s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl-propanoic acid dihydrate | |
(2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid;dihydrate |
Specifications
185°C (decomposition) | |
C9H13I2NO5 | |
3,5-diiodo-l-tyrosine dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, 3,5-diiodotyrosine dihydrate, 3,5-dijodtyrosindihydrat, c9h9i2no3.2h2o, 3,5-diiodo-l-tyrosine hydrate, l-tyrosine,3,5-diiodo-, hydrate 1:2, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoicaciddihydrate, 2s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl-propanoic acid dihydrate | |
O.O.N[C@@H](CC1=CC(I)=C(O)C(I)=C1)C(O)=O | |
469.01 | |
432.98 (Anhydrous) | |
May contain up to ≈10% water | |
(dry wt.) |
18835-59-1 | |
MFCD00150275 | |
YWAGQOOMOOUEGY-QVGJJETENA-N | |
(2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid;dihydrate | |
2724330 | |
98% | |
Light sensitive | |
3,5-Diiodo-L-tyrosine hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 206-092-0
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only