missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-L-tyrosine Dihydrate, TRC

Product Code. 30369534
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369534 500 mg -
30382354 2.5 g 2.5g
30373134 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369534 Supplier Toronto Research Chemicals Supplier No. TRCD466058500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18835-59-1
Molecular Formula C9H9I2NO3 . 2H2O
Molecular Weight (g/mol) 469.01
Synonym 3,5-Diiodo-L-tyrosine Dihydrate
SMILES O.O.N[C@@H](Cc1cc(I)c(O)c(I)c1)C(=O)O

Specifications

InChI Formula InChI=1S/C9H9I2NO3.2H2O/c10-5-1-4(2-6(11)8(5)13)3-7(12)9(14)15;;/h1-2,7,13H,3,12H2,(H,14,15);2*1H2/t7-;;/m0../s1
Quantity 500 mg
Formula Weight 468.8883
Shipping Condition Room Temperature
Chemical Name or Material 3,5-Diiodo-L-tyrosine Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.