missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-L-tyrosine dihydrate, 99%, Thermo Scientific™

Product Code. 15575056 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15575056 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15575056 Supplier Thermo Scientific Acros Supplier No. 225610050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 18835-59-1
Melting Point 200.0°C
Infrared Spectrum Authentic
Molecular Formula C9H13I2NO5
Linear Formula HOC6H2I2CH2CH(NH2)COOH·2H2O
MDL Number MFCD00150275
Quantity 5 g
Beilstein 14,IV,237
Specific Rotation Condition −20.70 (20.00°C c=2.5, 0.1N NaOH/H2O)
Specific Rotation -20.7
Synonym 3,5-diiodo-l-tyrosine dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, 3,5-diiodotyrosine dihydrate, 3,5-dijodtyrosindihydrat, c9h9i2no3.2h2o, 3,5-diiodo-l-tyrosine hydrate, l-tyrosine,3,5-diiodo-, hydrate 1:2, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoicaciddihydrate, 2s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl-propanoic acid dihydrate
InChI Key YWAGQOOMOOUEGY-QVGJJETENA-N
SMILES O.O.N[C@@H](CC1=CC(I)=C(O)C(I)=C1)C(O)=O
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid;dihydrate
Molecular Weight (g/mol) 469.01
PubChem CID 2724330
Formula Weight 469.01
Percent Purity 99%
Chemical Name or Material 3,5-Diiodo-L-tyrosine dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Diiodo-L-tyrosine dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.