missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-L-thyronine, TRC

Product Code. 17948514
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17948514 5 g 5g
17909094 500 mg 500mg
17908684 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17948514 Supplier Toronto Research Chemicals Supplier No. TRCD4551555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1041-01-6
Molecular Formula C15 H13 I2 N O4
Molecular Weight (g/mol) 525.08
Synonym L-Tyrosine, O-(4-hydroxyphenyl)-3,5-diiodo-, Alanine, 3-[4-(p-hydroxyphenoxy)-3,5-diiodophenyl]-, L- (8CI), 3,5-Diiodo-L-thyronine, 3,5-Diiodothyronine, Diiodo-L-thyronine, Diiodothyronine, L-3,5-Diiodothyronine, L-Diiodothyronine, L-T2, NSC 90469, O-(4-Hydroxyphenyl)-3,5-diiodo-L-tyrosine, Liothyronine Sodium Imp. E (EP)
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic acid
SMILES N[C@@H](Cc1cc(I)c(Oc2ccc(O)cc2)c(I)c1)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H13I2NO4/c16-11-5-8(7-13(18)15(20)21)6-12(17)14(11)22-10-3-1-9(19)2-4-10/h1-6,13,19H,7,18H2,(H,20,21)/t13-/m0/s1
Formula Weight 524.8934
Percent Purity >95%
Chemical Name or Material (2S)-2-Amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic Acid (3,5-Diiodo-L-thyronine)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.