missing translation for 'onlineSavingsMsg'
Learn More

3’,5’-Diiodo-D-thyronine, TRC

Product Code. 30373212
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373212 10 mg 10mg
30388244 1 mg 1mg
30386074 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30373212 Supplier Toronto Research Chemicals Supplier No. TRCD45515810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3',5'-Diiodo-D-thyronine
Analyte or Component Names 3',5'-Diiodo-D-thyronine
Shipping Condition Room Temperature
Molecular Formula C15 H13 I2 N O4
Synonym (R)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)phenyl)propanoic Acid, O-(4-Hydroxy-3,5-diiodophenyl)-D-tyrosine, 3-[p-(4-Hydroxy-3,5-diiodophenoxy)phenyl]-D-alanine
InChI Formula InChI=1S/C15H13I2NO4/c16-11-6-10(7-12(17)14(11)19)22-9-3-1-8(2-4-9)5-13(18)15(20)21/h1-4,6-7,13,19H,5,18H2,(H,20,21)/t13-/m1/s1
Solution Type Neat
SMILES N[C@H](Cc1ccc(Oc2cc(I)c(O)c(I)c2)cc1)C(=O)O
IUPAC Name (2R)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)phenyl]propanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 525.077
Quantity 10 mg
Formula Weight 524.893
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.