missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-4-hydroxybenzoic Acid, Mikromol™

Product Code. 30052490
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052490 100 mg 100mg
30052114 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052490 Supplier Mikromol Supplier No. MM0694.14
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3,5-Diiodo-4-hydroxybenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Diiodo-4-hydroxybenzoic Acid
CAS 618-76-8
Analyte or Component Names 3,5-Diiodo-4-hydroxybenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C7 H4 I2 O3
Certifications/Compliance ISO 9001
Synonym 3,5-Diiodo-4-hydroxybenzoic Acid, Benzoic acid, 4-hydroxy-3,5-diiodo-, 3,5-Diiodo-4-hydroxybenzoic acid, 4-Hydroxy-3,5-diiodobenzoic acid, Ioxynil acid, M and B 10903, NSC 1497
InChI Formula InChI=1S/C7H4I2O3/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1-2,10H,(H,11,12)
SMILES OC(=O)c1cc(I)c(O)c(I)c1
IUPAC Name 4-hydroxy-3,5-diiodobenzoic acid
Molecular Weight (g/mol) 389.91
Quantity 100 mg
Formula Weight 389.825 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.