missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxyphenylpropanoic Acid 3-O-b-D-Glucuronide(>85%), TRC

Product Code. 30385440
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385440 2.5 mg 2.5mg
30387826 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385440 Supplier Toronto Research Chemicals Supplier No. TRCD4543702.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Dihydroxyphenylpropanoic Acid 3-O-beta-D-Glucuronide
Analyte or Component Names 3,5-Dihydroxyphenylpropanoic Acid 3-O-beta-D-Glucuronide
Shipping Condition Room Temperature
Molecular Formula C15H18O10
Synonym 3-(3,5-Dihydroxyphenyl)-1-propanoic Acid β-D-Glucuronide, DHPPA β-D-Glucuronide
InChI Formula InChI=1S/C15H18O10/c16-7-3-6(1-2-9(17)18)4-8(5-7)24-15-12(21)10(19)11(20)13(25-15)14(22)23/h3-5,10-13,15-16,19-21H,1-2H2,(H,17,18)(H,22,23)/t10-,11-,12+,13-,15+/m0/s1
Solution Type Neat
SMILES OC1=CC(CCC(O)=O)=CC(O[C@@H]2O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]2O)=C1
Industry Type Forensics
Molecular Weight (g/mol) 358.3
Quantity 2.5 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.