missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzyl alcohol, 98%, Thermo Scientific™

Product Code. p-7028110
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410788 5 g 5g
11420788 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410788 Supplier Thermo Scientific Alfa Aesar Supplier No. B25248.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dihydroxybenzyl alcohol may be used as monomer for the synthesis of a series of monodisperse dendritic polyether macromolecules. It may be employed in the synthesis of dimethylsilyl linked dihydroxybenzyl alcohol based dendrimers.

Solubility
Soluble in DMSO, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29654-55-5
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00016867
InChI Key NGYYFWGABVVEPL-UHFFFAOYSA-N
Synonym 3,5-dihydroxybenzyl alcohol, 5-hydroxymethyl-1,3-benzenediol, 5-hydroxymethyl benzene-1,3-diol, 5-hydroxymethyl resorcinol, benzyl alcohol, 3,5-dihydroxy, 1,3-benzenediol, 5-hydroxymethyl, 3,5-dihydroxybenzylalcohol, 3,5-dihydroxy benzyl alcohol, pubchem7418, acmc-1cjog
PubChem CID 34661
IUPAC Name 5-(hydroxymethyl)benzene-1,3-diol
SMILES C1=C(C=C(C=C1O)O)CO

Specifications

Melting Point 182°C to 186°C (decomposition)
Quantity 5 g
Solubility Information Soluble in DMSO,methanol.
Formula Weight 140.14
Percent Purity 98%
Chemical Name or Material 3,5-Dihydroxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.