missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzonitrile, 98%

Product Code. 11337525
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11337525

Brand: Thermo Scientific Alfa Aesar L05642.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19179-36-3 | C7H5NO2 | 135.12 g/mol

3,5-Dihydroxybenzonitrile is used in the preparation of 3-hydroxy-5-methoxybenzonitrile using potassium carbonate as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dihydroxybenzonitrile is used in the preparation of 3-hydroxy-5-methoxybenzonitrile using potassium carbonate as a reagent.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19179-36-3
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
MDL Number MFCD00016453
InChI Key ABHOEQJNEOMTEK-UHFFFAOYSA-N
Synonym benzonitrile, 3,5-dihydroxy, benzonitrile,3,5-dihydroxy, 3,5-dihydroxy benzonitrile, 5-cyanoresorcinol, a-resorcylonitrile, pubchem13116, acmc-1bouo
PubChem CID 87952
IUPAC Name 3,5-dihydroxybenzonitrile
SMILES OC1=CC(=CC(O)=C1)C#N

Specifications

Melting Point 185°C to 190°C
Quantity 5 g
UN Number UN3439
Beilstein 6322491
Solubility Information Soluble in water.
Formula Weight 135.12
Percent Purity 98%
Chemical Name or Material 3,5-Dihydroxybenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.