missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzoic Acid Methyl Ester, TRC

Product Code. 30380290
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30380290 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380290 Supplier Toronto Research Chemicals Supplier No. TRCD45171510G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Dihydroxybenzoic Acid Methyl Ester
CAS 2150-44-9
Molecular Formula C8 H8 O4
Quantity 10 g
Synonym Benzoic acid, 3,5-dihydroxy-, methyl ester, α-Resorcylic acid, methyl ester (6CI,7CI,8CI), 3,5-Dihydroxybenzoic acid methyl ester, Methyl 3,5-dihydroxybenzoate, Methyl 3,5-hydroxybenzoate, Methyl α-resorcylate, NSC 61082, α-Resorcinol carboxylic acid methyl ester
InChI Formula InChI=1S/C8H8O4/c1-12-8(11)5-2-6(9)4-7(10)3-5/h2-4,9-10H,1H3
SMILES COC(=O)c1cc(O)cc(O)c1
IUPAC Name methyl 3,5-dihydroxybenzoate
Molecular Weight (g/mol) 168.15
Formula Weight 168.0423
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.