missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzoic acid, 97%

Product Code. p-112357 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10122860

Brand: Thermo Scientific Acros 114901000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-10-5 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-10-5
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00002512
InChI Key UYEMGAFJOZZIFP-UHFFFAOYSA-N
Synonym alpha-resorcylic acid, 5-carboxyresorcinol, benzoic acid, 3,5-dihydroxy, unii-2wc5lmo6l1, 3,5-dihydroxy benzoic acid, 3,5-dihydroxy-benzoic acid, 3,5-dhba, 3,5-dihydroxybenzoicacid, a-resorcylic acid, .alpha.-resorcylic acid
PubChem CID 7424
ChEBI CHEBI:39912
IUPAC Name 3,5-dihydroxybenzoic acid
SMILES OC(=O)C1=CC(O)=CC(O)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 235.0°C to 240.0°C
Color White to Beige
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula (HO)2C6H3CO2H
Quantity 100 g
Beilstein 10,404
Solubility Information Solubility in water: 84g/L (20°C). Other solubilities: soluble in ethanol and ether
Formula Weight 154.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3,5-Dihydroxybenzoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dihydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.