missing translation for 'onlineSavingsMsg'
Learn More

3’,5’-Dihydrodiol Simvastatin(Mixture of Diastereomers), TRC

Product Code. 30388230
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388230 100 mg
30373124 50 mg
30376616 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388230 Supplier Toronto Research Chemicals Supplier No. TRCD452690100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198818-02-9
Molecular Formula C25 H40 O7
Molecular Weight (g/mol) 452.58
Synonym [1S-[1α,3α,4β,6α,7β,8β(2S*,4S*),8aβ]]-Butanoic Acid 2,2-Dimethyl-1,2,3,4,6,7,8,8a- octahydro-4,6-dihydroxy-3,7-dimethyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl) ethyl]-1-naphthalenyl Ester
SMILES O=C(C(C)(C)CC)O[C@H]1C[C@@H](C)C(O)C2=CC(O)[C@H](C)[C@H](CC[C@H]3OC(C[C@H](O)C3)=O)[C@]21[H]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C25H40O7/c1-6-25(4,5)24(30)32-20-9-13(2)23(29)18-12-19(27)14(3)17(22(18)20)8-7-16-10-15(26)11-21(28)31-16/h12-17,19-20,22-23,26-27,29H,6-11H2,1-5H3/t13-,14-,15-,16-,17+,19?,20+,22-,23?/m1/s1
Percent Purity >95%
Chemical Name or Material 3',5'-Dihydrodiol Simvastatin (Mixture of Diastereomers)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.