missing translation for 'onlineSavingsMsg'
Learn More

3,5-dihydro-2H-1,5-benzothiazepin-4-one, TRC

Product Code. 30384165
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384165 250 mg
30373607 50 mg
30377715 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384165 Supplier Toronto Research Chemicals Supplier No. TRCD457148250MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53454-43-6
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.24
Synonym 2,3,4,5-Tetrahydro-1,5-benzothiazepin-4-one
IUPAC Name 3,5-dihydro-2H-1,5-benzothiazepin-4-one
SMILES O=C1CCSc2ccccc2N1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H9NOS/c11-9-5-6-12-8-4-2-1-3-7(8)10-9/h1-4H,5-6H2,(H,10,11)
Formula Weight 179.0405
Chemical Name or Material 3,5-dihydro-2H-1,5-benzothiazepin-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.