missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorosalicylic acid, 98+%, Thermo Scientific Chemicals

Product Code. 15445727
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445727 1 g 1g
15455727 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445727 Supplier Thermo Scientific Alfa Aesar Supplier No. H32940.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84376-20-5
Molecular Formula C7H4F2O3
Molecular Weight (g/mol) 174.103
MDL Number MFCD06203609
InChI Key GZPCNALAXFNOBT-UHFFFAOYSA-N
Synonym 3,5-difluorosalicylic acid, 3,5-difluoro-salicylic acid, benzoic acid, 3,5-difluoro-2-hydroxy
PubChem CID 10888405
IUPAC Name 3,5-difluoro-2-hydroxybenzoic acid
SMILES C1=C(C=C(C(=C1F)O)C(=O)O)F

Specifications

Melting Point 187°C to 191°C
Quantity 1 g
Formula Weight 174.1
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluorosalicylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.