missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluorophenylhydrazine hydrochloride, 97+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 97.0 |
CAS Max % | 100.0 |
Melting Point | 261.0°C to 266.0°C |
Assay Percent Range | 97% min. (HPLC) |
Packaging | Glass bottle |
Linear Formula | F2C6H3NHNH2·HCl |
Quantity | 1 g |
Formula Weight | 180.58 |
Percent Purity | 97+% |
Chemical Name or Material | 3, 5-Difluorophenylhydrazine hydrochloride |
Safety and Handling
Product Identifier
- 3,5-Difluorophenylhydrazine hydrochloride
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'