missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluorobenzyl alcohol, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3,5-Difluorobenzyl alcohol is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
3,5-Difluorobenzyl alcohol is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Density | 1.276 |
Boiling Point | 98°C to 99°C (9 mmHg) |
Flash Point | 94°C (201°F) |
Refractive Index | 1.487 |
Quantity | 5 g |
Beilstein | 5499788 |
Synonym | 3,5-difluorobenzyl alcohol, 3,5-difluorophenyl methanol, benzenemethanol, 3,5-difluoro, 3,5-difluorophenyl methan-1-ol, 3,5-difluorobenzylalcohol, 3, 5-difluorobenzyl alcohol, pubchem4918, acmc-1baxh, ksc493g5j, 3,5-difluoro-phenyl methanol |
Formula Weight | 144.12 |
Percent Purity | 97% |
Chemical Name or Material | 3,5-Difluorobenzyl alcohol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'