missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzoyl chloride, 98+%

Product Code. 11419357
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11419357

Brand: Thermo Scientific Alfa Aesar B21400.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 129714-97-2 | C7H3ClF2O | 176.547 g/mol

3,5-Difluorobenzoyl chloride, is used as an pharmaceutical intermediate, organic intermediate and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluorobenzoyl chloride, is used as an pharmaceutical intermediate, organic intermediate and chemical synthesis.

Solubility
It reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids and bases. Moisture Sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129714-97-2
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00010309
InChI Key OYZWEOORLJBPMA-UHFFFAOYSA-N
Synonym 3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride
PubChem CID 145600
IUPAC Name 3,5-difluorobenzoyl chloride
SMILES C1=C(C=C(C=C1F)F)C(=O)Cl

Specifications

Density 1.41
Boiling Point 173°C to 175°C
Flash Point 69°C (156°F)
Refractive Index 1.503
Quantity 5 g
UN Number UN3265
Beilstein 4307685
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 176.55
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.