missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluorobenzeneboronic acid neopentyl glycol ester, 98+%

Product Code. 11394006
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394006 1 g 1g
11304016 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394006 Supplier Alfa Aesar™ Supplier No. L17266.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 216393-57-6
Molecular Formula C11H13BF2O2
Molecular Weight (g/mol) 226.03
MDL Number MFCD01318139
InChI Key OLHRAEPCGZMMTI-UHFFFAOYSA-N
Synonym 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborinane, 3,5-difluorobenzeneboronic acid neopentyl glycol ester, 3,5-difluorophenylboronic acid neopentyl glycol ester, 3,5-difluorobenzeneboronic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborane, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborine, 1,3,2-dioxaborinane, 2-3,5-difluorophenyl-5,5-dimethyl, 3,5-difluorobenzenebornoic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyldihydro-4h-1,3,2-dioxaborin
PubChem CID 2774056
IUPAC Name 2-(3,5-difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=CC(=CC(=C2)F)F

Specifications

Melting Point 43°C to 45°C
Quantity 1 g
Formula Weight 226.03
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluorobenzeneboronic acid neopentyl glycol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.