missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzaldehyde, 98+%

Product Code. 10433405 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10433405 5 g Glass bottle 5g
10117893 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10433405 Supplier Thermo Scientific Acros Supplier No. 293470050
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32085-88-4 | C7H4F2O | 142.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32085-88-4
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.1
MDL Number MFCD00010329
InChI Key ASOFZHSTJHGQDT-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-difluoro, 3,5-difluoro-benzaldehyde, pubchem1414, 3,5difluorobenzaldehyde, acmc-1aewe, 3,5-diflourobenzaldehyde, 3-5-difluorobenzaldehyde, 3, 5-difluorobenzaldehyde, ksc224o7p, 3,5-difluorophenyl methanone
PubChem CID 588160
IUPAC Name 3,5-difluorobenzaldehyde
SMILES C1=C(C=C(C=C1F)F)C=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Green
Density 1.2960g/mL
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3CHO
Refractive Index 1.4930 to 1.4950
Quantity 5 g
Specific Gravity 1.296
Formula Weight 142.1
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 3, 5-Difluorobenzaldehyde, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,5-Difluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.