missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Difluoro-4'-methoxyacetophenone, 97%, Thermo Scientific™

Product Code. 11366979
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11366979 5 g 5g
11356979 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366979 Supplier Thermo Scientific Alfa Aesar Supplier No. H26833.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3′,5′-Difluoro-4′-methoxyacetophenone, is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170570-79-3
Molecular Formula C9H8F2O2
Molecular Weight (g/mol) 186.158
MDL Number MFCD00236243
InChI Key OUJZNFJFMAKGMS-UHFFFAOYSA-N
Synonym 1-3,5-difluoro-4-methoxyphenyl ethanone, 3',5'-difluoro-4'-methoxyacetophenone, 3,5-difluoro-4-methoxyacetophenone, 1-3,5-difluoro-4-methoxyphenyl ethan-1-one, 1-3,5-difluoro-4-methoxy-phenyl ethanone, 4-acetyl-2,6-difluoroanisole, 3.5-difluoro-4-methoxyacetophenone, 1-acetyl-3,5-difluoro-4-methoxybenzene, 3',5'-difluoro-4-difluoro-4-methoxyacetophenone-methoxyacetophenone, 1-3,5-difluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-2,6-difluoroanisole
PubChem CID 2736983
IUPAC Name 1-(3,5-difluoro-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C(=C1)F)OC)F

Specifications

Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 186.15
Percent Purity 97%
Chemical Name or Material 3',5'-Difluoro-4'-methoxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.