missing translation for 'onlineSavingsMsg'
Learn More

(3,5-Difluoro-2-methoxyphenyl)boronic acid, 97%, Thermo Scientific™

Product Code. 10423175 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10423175 5 g Glass bottle 5g
10743983 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423175 Supplier Thermo Scientific Acros Supplier No. 365820050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kemiske identifikatorer

CAS 737000-76-9
Molecular Formula C7H7BF2O3
Molecular Weight (g/mol) 187.94
MDL Number MFCD03095353
InChI Key JXQHRWOUVJRCSR-UHFFFAOYSA-N
Synonym 3,5-difluoro-2-methoxyphenyl boronic acid, 3,5-difluoro-2-methoxybenzeneboronic acid, 2-methoxy-3,5-difluorophenylboronic acid, acmc-209orz, 2-methoxy-3,5-difluoro-benzeneboronic acid, boronic acid, 3,5-difluoro-2-methoxyphenyl, boronic acid,b-3,5-difluoro-2-methoxyphenyl, 3,5-difluoro-2-methoxyphenyl boronicacid, 3,5-bis fluoranyl-2-methoxy-phenyl boronic acid, b-3,5-difluoro-2-methoxyphenyl-boronic acid
PubChem CID 2734607
IUPAC Name (3,5-difluoro-2-methoxyphenyl)boronic acid
SMILES COC1=C(F)C=C(F)C=C1B(O)O

Tekniske data

CAS Min % 96.0
CAS Max % 100.0
Melting Point 200°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 187.81
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material (3, 5-Difluoro-2-methoxyphenyl)boronic acid, 97%
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • (3,5-Difluoro-2-methoxyphenyl)boronic acid
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.