missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-2-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11349725
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 737000-76-9
Molecular Formula C7H7BF2O3
Molecular Weight (g/mol) 187.94
MDL Number MFCD03095353
InChI Key JXQHRWOUVJRCSR-UHFFFAOYSA-N
Synonym 3,5-difluoro-2-methoxyphenyl boronic acid, 3,5-difluoro-2-methoxybenzeneboronic acid, 2-methoxy-3,5-difluorophenylboronic acid, acmc-209orz, 2-methoxy-3,5-difluoro-benzeneboronic acid, boronic acid, 3,5-difluoro-2-methoxyphenyl, boronic acid,b-3,5-difluoro-2-methoxyphenyl, 3,5-difluoro-2-methoxyphenyl boronicacid, 3,5-bis fluoranyl-2-methoxy-phenyl boronic acid, b-3,5-difluoro-2-methoxyphenyl-boronic acid
PubChem CID 2734607
IUPAC Name (3,5-difluoro-2-methoxyphenyl)boronic acid
SMILES COC1=C(F)C=C(F)C=C1B(O)O

Specifications

Melting Point 112°C to 115°C
Quantity 5 g
Formula Weight 187.94
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-2-methoxybenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'