missing translation for 'onlineSavingsMsg'
Learn More

3,5-Didesamino-3,5-dioxo Lamotrigine, TRC

Product Code. 30383840
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383840 50 mg
30373865 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383840 Supplier Toronto Research Chemicals Supplier No. TRCD44040050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 661463-79-2
Molecular Formula C9 H5 Cl2 N3 O2
Molecular Weight (g/mol) 258.06
Synonym 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5(2H,4H)-dione, Lamotrigine Imp. D (EP)
IUPAC Name 6-(2,3-dichlorophenyl)-2H-1,2,4-triazine-3,5-dione
SMILES Clc1cccc(C2=NNC(=O)NC2=O)c1Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H5Cl2N3O2/c10-5-3-1-2-4(6(5)11)7-8(15)12-9(16)14-13-7/h1-3H,(H2,12,14,15,16)
Formula Weight 256.9759
Chemical Name or Material 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5(2H,4H)-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.