missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenylhydrazine hydrochloride, 95%

Product Code. 11412098
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412098 5 g 5g
11402098 1 g 1g
11422098 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412098 Supplier Thermo Scientific Alfa Aesar Supplier No. A15127.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63352-99-8 | C6H7Cl3N2 | 213.486 g/mol

3,5-Dichlorophenyl)hydrazine Hydrochloride is a building block that has been used for the synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives to be used as and anti-leukemic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorophenyl)hydrazine Hydrochloride is a building block that has been used for the synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives to be used as and anti-leukemic agent.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63352-99-8
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number RONG>
InChI Key BDXZOJVMTJOAPS-UHFFFAOYSA-N
Synonym 3,5-dichlorophenylhydrazine hydrochloride, 3,5-dichlorophenyl hydrazine hydrochloride, 3,5-dichlorophenyl hydrazine monohydrochloride, 3,5-dichlorophenylhydrazine hcl, 1-3,5-dichlorophenyl hydrazine hydrochloride, hydrazine, 3,5-dichlorophenyl-, monohydrochloride, 3,5-dichlorophenylhydrazinehydrochloride, pubchem7531, c6h6cl2n2.hcl, acmc-209nf3
PubChem CID 2723913
IUPAC Name (3,5-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=C(C=C(C=C1Cl)Cl)NN.Cl

Specifications

Melting Point ∼204°C (decomposition)
Quantity 5 g
Beilstein 4208459
Solubility Information Soluble in water.
Formula Weight 213.5
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorophenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.