Learn More
(3,5-Dichlorophenyl)methanesulfonyl chloride, 97%, Thermo Scientific Chemicals
£211.00 - £777.57
Chemical Identifiers
CAS | 163295-70-3 |
---|---|
Molecular Formula | C7H5Cl3O2S |
Molecular Weight (g/mol) | 259.53 |
MDL Number | MFCD01631928 |
InChI Key | GZMJRPMBVICCFL-UHFFFAOYSA-N |
Synonym | 3,5-dichlorophenyl methanesulfonyl chloride, 3,5-dichloro-phenyl-methanesulfonyl chloride, acmc-1c80o, 3,5-dichloro-, a-toluenesufonyl chloride, 3,5-dichlorophenyl methyl chlorosulfone, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichloro-alpha-toluenesulfonyl chloride, 3,5-dichlorophenyl methane-sulfonyl chloride, benzenemethanesulfonylchloride, 3,5-dichloro, 3,5-dichlorophenyl methyl sulfonyl chloride |
PubChem CID | 4993270 |
IUPAC Name | (3,5-dichlorophenyl)methanesulfonyl chloride |
SMILES | ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
163295-70-3 | |
259.53 | |
GZMJRPMBVICCFL-UHFFFAOYSA-N | |
4993270 | |
ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1 |
C7H5Cl3O2S | |
MFCD01631928 | |
3,5-dichlorophenyl methanesulfonyl chloride, 3,5-dichloro-phenyl-methanesulfonyl chloride, acmc-1c80o, 3,5-dichloro-, a-toluenesufonyl chloride, 3,5-dichlorophenyl methyl chlorosulfone, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichloro-alpha-toluenesulfonyl chloride, 3,5-dichlorophenyl methane-sulfonyl chloride, benzenemethanesulfonylchloride, 3,5-dichloro, 3,5-dichlorophenyl methyl sulfonyl chloride | |
(3,5-dichlorophenyl)methanesulfonyl chloride |
Specifications
163295-70-3 | |
100.0 | |
White to Yellow | |
96% min. as diethylsulfonamide derivative (HPLC) | |
MFCD01631928 | |
GZMJRPMBVICCFL-UHFFFAOYSA-N | |
(3,5-dichlorophenyl)methanesulfonyl chloride | |
4993270 | |
97% | |
(3,5-Dichlorophenyl)methanesulfonyl chloride |
96.0 | |
108.6°C to 110.6°C | |
Authentic | |
C7H5Cl3O2S | |
3,5-dichlorophenyl methanesulfonyl chloride, 3,5-dichloro-phenyl-methanesulfonyl chloride, acmc-1c80o, 3,5-dichloro-, a-toluenesufonyl chloride, 3,5-dichlorophenyl methyl chlorosulfone, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichloro-alpha-toluenesulfonyl chloride, 3,5-dichlorophenyl methane-sulfonyl chloride, benzenemethanesulfonylchloride, 3,5-dichloro, 3,5-dichlorophenyl methyl sulfonyl chloride | |
ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1 | |
259.53 | |
259.54 | |
Powder or Solid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- (3,5-Dichlorophenyl) methanesulfonyl chloride, 97%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only