missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenol, 98%

Product Code. 11319529
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
This item is not returnable. View return policy

Product Code. 11319529

Brand: Thermo Scientific Alfa Aesar A15328.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-35-5 | C6H4Cl2O | 162.997 g/mol

3,5-Dichlorophenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorophenol is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water. Very soluble in ethanol, ethyl ether and petroleum ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-35-5
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 162.997
MDL Number MFCD00002259
InChI Key VPOMSPZBQMDLTM-UHFFFAOYSA-N
Synonym phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
PubChem CID 11571
IUPAC Name 3,5-dichlorophenol
SMILES C1=C(C=C(C=C1Cl)Cl)O

Specifications

Melting Point 63°C to 68°C
Boiling Point 233°C
Odor Phenol-like
Quantity 25 g
UN Number UN2020
Beilstein 1906458
Solubility Information Slightly soluble in water. Very soluble in ethanol,ethyl ether and petroleum ether.
Formula Weight 163
Percent Purity 98%
Chemical Name or Material 3,5-Dichlorophenol
compliance-icons
Product Identifier
  • 3,5-Dichlorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 3,5-Dichlorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.