missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloroisonicotinic acid, 98%, Thermo Scientific™

Product Code. 10316684 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10316684 1 g Glass bottle 1g
10326684 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10316684 Supplier Thermo Scientific Acros Supplier No. 399020010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13958-93-5
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192
InChI Key BUQPTOSHKHYHHB-UHFFFAOYSA-N
Synonym 3,5-dichloroisonicotinic acid, 3,5-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3,5-dichloro, dichloroisonicotinicacid, acmc-1cbzu, 3?5-dichloroisonicotinic acid, 3,5-dichloro-isonicotinic acid, buqptoshkhyhhb-uhfffaoysa, 3,5-dichloroisonicotinic acid #, 3,5-dichloro-pyridine-4-carboxylic acid
PubChem CID 605763
IUPAC Name 3,5-dichloropyridine-4-carboxylic acid
SMILES C1=C(C(=C(C=N1)Cl)C(=O)O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 230.0°C
Assay Percent Range 97.5% min. (Acid)
Packaging Glass bottle
Quantity 1 g
Formula Weight 192
Percent Purity 98%
Chemical Name or Material 3, 5-Dichloroisonicotinic acid
compliance-icons
Product Identifier
  • 3,5-Dichloroisonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.