missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzylamine, 97%, Thermo Scientific™

Product Code. 15322052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15322052 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15322052 Supplier Thermo Scientific Acros Supplier No. 454850010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39989-43-0
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00052681
InChI Key ICIJWOWQUHHETJ-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzylamine, 3,5-dichlorophenyl methanamine, benzenemethanamine, 3,5-dichloro, 3,5-dichloro-benzylamine, 1-3,5-dichlorophenyl methanamine, 3,5-dichlorophenyl methylamine, pubchem23431, 3.5-dichlorobenzylamine, acmc-1cthv, integrase inhibitor, r1 4
PubChem CID 457602
SMILES NCC1=CC(Cl)=CC(Cl)=C1

Specifications

Density 1.3200g/mL
Boiling Point 116.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Linear Formula Cl2C6H3CH2NH2
Refractive Index 1.575
Quantity 1 g
Specific Gravity 1.32
IUPAC Name (3,5-dichlorophenyl)methanamine
Formula Weight 176.05
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorobenzylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.