missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzylamine, 95%, Thermo Scientific™

Catalog Number p-2698336
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
39989-43-0
C7H7Cl2N
176.04
MFCD00052681
ICIJWOWQUHHETJ-UHFFFAOYSA-N
3,5-dichlorobenzylamine, 3,5-dichlorophenyl methanamine, benzenemethanamine, 3,5-dichloro, 3,5-dichloro-benzylamine, 1-3,5-dichlorophenyl methanamine, 3,5-dichlorophenyl methylamine, pubchem23431, 3.5-dichlorobenzylamine, acmc-1cthv, integrase inhibitor, r1 4
457602
NCC1=CC(Cl)=CC(Cl)=C1
This item is not returnable. View return policy
39989-43-0
C7H7Cl2N
176.04
MFCD00052681
ICIJWOWQUHHETJ-UHFFFAOYSA-N
3,5-dichlorobenzylamine, 3,5-dichlorophenyl methanamine, benzenemethanamine, 3,5-dichloro, 3,5-dichloro-benzylamine, 1-3,5-dichlorophenyl methanamine, 3,5-dichlorophenyl methylamine, pubchem23431, 3.5-dichlorobenzylamine, acmc-1cthv, integrase inhibitor, r1 4
457602
NCC1=CC(Cl)=CC(Cl)=C1
CAS Min % 95.0
Density 1.3200g/mL
Assay Percent Range 95%
Linear Formula Cl2C6H3CH2NH2
Quantity 1 g
IUPAC Name (3,5-dichlorophenyl)methanamine
Formula Weight 176.05
Percent Purity 95%
Chemical Name or Material 3, 5-Dichlorobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'