Learn More
3,5-Dichlorobenzyl alcohol, 98%, ACROS Organics™
Brand: Acros Organics 182770050
Code : 29
Additional Details : CAS Number : 60211-57-6 Weight : 0.06000kg
Quantity | 5g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
3, 5-Dichlorobenzyl alcohol | |
60211-57-6 | |
100.0 | |
98% | |
Cl2C6H3CH2OH | |
MFCD00004634 |
98% | |
97.5 | |
≥97.5% | |
C7H6Cl2O | |
177.03 |
Specifications
3, 5-Dichlorobenzyl alcohol | |
76° to 80°C | |
98% | |
97.5 | |
Authentic | |
Crystalline Powder | |
98% | |
Cl2C6H3CH2OH | |
MFCD00004634 |
White to light yellow | |
5g | |
60211-57-6 | |
100.0 | |
Glass bottle | |
≥97.5% | |
C7H6Cl2O | |
177.03 |