missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzoyl chloride, 96%

Product Code. 11470538
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11470538

Brand: Thermo Scientific Alfa Aesar B24676.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2905-62-6 | C7H3Cl3O | 209.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-62-6
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00009817
InChI Key GGHLXLVPNZMBQR-UHFFFAOYSA-N
Synonym benzoyl chloride, 3,5-dichloro, 3,5-dichlorobenzoylchloride, dichlorobenzoylchloride 3,5-, pubchem10805, dsstox_cid_7506, acmc-209h6l, dsstox_rid_78478, dsstox_gsid_27506, ksc204k6p, 3,5-dichloro benzoyl chloride
PubChem CID 76191
IUPAC Name 3,5-dichlorobenzoyl chloride
SMILES ClC(=O)C1=CC(Cl)=CC(Cl)=C1

Specifications

Melting Point 24°C to 28°C
Density 1.478
Boiling Point 245°C
Flash Point 137°C (278°F)
Refractive Index 1.582
Quantity 100 g
UN Number UN3261
Beilstein 1940681
Sensitivity Moisture sensitive
Formula Weight 209.46
Percent Purity 96%
Chemical Name or Material 3,5-Dichlorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.