missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzotrifluoride, 98%, Thermo Scientific™

Product Code. 10125560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10125560 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10125560 Supplier Thermo Scientific Acros Supplier No. 299600050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 54773-20-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point -13°C to 12°C
Color Colorless to Yellow
Density 1.4700g/mL
Boiling Point 173°C to 174°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H3Cl2F3
Refractive Index 1.4710 to 1.473
MDL Number MFCD00203929
Quantity 5 g
Specific Gravity 1.47
Synonym 3,5-dichlorobenzotrifluoride, 1,3-dichloro-5-trifluoromethyl benzene, benzene, 1,3-dichloro-5-trifluoromethyl, 1,5-dichloro-3-trifluoromethyl benzene, pubchem19726, ksc949c6r, 3,5-dichlorobenzotrifluoride,98, 1-trifluoromethyl-3,5-dichlorobenzene, 3,5-dichloro-a,a,a-trifluorotoluene, 3 5-dichlorobenzotrifluoride
InChI Key PCMPDUPKYRRIIP-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1Cl)Cl)C(F)(F)F
IUPAC Name 1,3-dichloro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol) 215
PubChem CID 35361
Formula Weight 215
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 5-Dichlorobenzotrifluoride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.