missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzhydrazide, 97%

Product Code. 11450487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450487 1 g 1g
11460487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450487 Supplier Thermo Scientific Alfa Aesar Supplier No. B22383.03
 Restricted Item
Estimated Shipment: 16-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 62899-78-9 | C7H6Cl2N2O | 205.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62899-78-9
Molecular Formula C7H6Cl2N2O
Molecular Weight (g/mol) 205.04
MDL Number MFCD00833411
InChI Key DNISXKBKTSHNKL-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzhydrazide, 3,5-dichlorobenzenecarbohydrazide, acmc-1b4zh, 3,5-bis chloranyl benzohydrazide, 3,5-dichlorobenzoic acid hydrazide, benzoic acid,3,5-dichloro-,hydrazide, benzoic acid,3,5-dichloro-, hydrazide
PubChem CID 2735966
IUPAC Name 3,5-dichlorobenzohydrazide
SMILES NNC(=O)C1=CC(Cl)=CC(Cl)=C1

Specifications

Melting Point 205°C to 207°C
Quantity 1 g
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorobenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.