missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzene-1-carboximidamide hydrochloride, Tech., Thermo Scientific™

Product Code. 10203532
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10203532 10 g Amber Glass Bottle 10g
10059591 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10203532 Supplier Thermo Scientific Maybridge Supplier No. SPB00407EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 22978-61-6
Molecular Formula C7H7Cl3N2
Molecular Weight (g/mol) 225.50
MDL Number MFCD00173785
InChI Key FFLPJEKRAYZAMU-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzimidamide hydrochloride, 3,5-dichlorobenzene-1-carboximidamide hydrochloride, 3,5-dichlorobenzenecarboximidamide hydrochloride, 3,5-dichlorobenzamidine hydrochloride, 3,5-dichloro-benzamidine hydrochloride, bestipharma 538-443, 3,5-dichlorobenzenecarboxamidine, chloride, 3,5-dichlorobenzene-1-carboximidamide hcl, 3,5-dichlorobenzene-1-carboximidamide hydrochlorid, 3,5-bis chloranyl benzenecarboximidamide hydrochloride
PubChem CID 2743159
SMILES Cl.NC(=N)C1=CC(Cl)=CC(Cl)=C1

Specifications

Packaging Amber Glass Bottle
Linear Formula Cl2C6H3C(=NH)NH2·HCl
Quantity 10 g
IUPAC Name 3,5-dichlorobenzene-1-carboximidamide hydrochloride
Formula Weight 225.51
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3, 5-Dichlorobenzene-1-carboximidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.