missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzene-1-carboximidamide hydrochloride, Tech., Thermo Scientific™

Product Code. p-2699484
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
Product Code. Quantity Packaging unitSize
10059591 1 g Amber Glass Bottle 1g
10203532 10 g Amber Glass Bottle 10g
2 options
This item is not returnable. View return policy

Product Code. 10059591

Brand: Thermo Scientific Maybridge SPB00407DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22978-61-6
Molecular Formula C7H7Cl3N2
Molecular Weight (g/mol) 225.50
MDL Number MFCD00173785
InChI Key FFLPJEKRAYZAMU-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzimidamide hydrochloride, 3,5-dichlorobenzene-1-carboximidamide hydrochloride, 3,5-dichlorobenzenecarboximidamide hydrochloride, 3,5-dichlorobenzamidine hydrochloride, 3,5-dichloro-benzamidine hydrochloride, bestipharma 538-443, 3,5-dichlorobenzenecarboxamidine, chloride, 3,5-dichlorobenzene-1-carboximidamide hcl, 3,5-dichlorobenzene-1-carboximidamide hydrochlorid, 3,5-bis chloranyl benzenecarboximidamide hydrochloride
PubChem CID 2743159
SMILES Cl.NC(=N)C1=CC(Cl)=CC(Cl)=C1

Specifications

Packaging Amber Glass Bottle
Linear Formula Cl2C6H3C(=NH)NH2·HCl
Quantity 1 g
IUPAC Name 3,5-dichlorobenzenecarboximidamide;hydrochloride
Formula Weight 225.51
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3, 5-Dichlorobenzene-1-carboximidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.