missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzenamine, TRC

Product Code. 30134401
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134401 100 g 100g
18021930 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134401 Supplier Toronto Research Chemicals Supplier No. TRCD431855100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-43-7
Molecular Formula C6 H5 Cl2 N
Molecular Weight (g/mol) 162.02
Synonym Aniline, 3,5-dichloro- (7CI,8CI), 3,5-Dichlorobenzenamine, 3,5-DCA, 3,5-Dichloroaniline, 3,5-Dichlorolaniline, 3,5-Dichlorophenylamine
IUPAC Name 3,5-dichloroaniline
SMILES Nc1cc(Cl)cc(Cl)c1

Specifications

Analyte or Component Names 3,5-Dichloroaniline
Shipping Condition Room Temperature
InChI Formula InChI=1S/C6H5Cl2N/c7-4-1-5(8)3-6(9)2-4/h1-3H,9H2
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 g
Formula Weight 160.9799
Chemical Name or Material 3,5-Dichloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.