missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzaldehyde, 97%

Product Code. 11460933
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460933 5 g 5g
11450933 1 g 1g
11470933 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460933 Supplier Thermo Scientific Alfa Aesar Supplier No. A13325.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10203-08-4 | C7H4Cl2O | 175.008 g/mol

3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.

Solubility
Insoluble in water.

Notes
Air sensitive. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10203-08-4
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003352
InChI Key CASRSOJWLARCRX-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dichloro, 3,5-dichloro-benzaldehyde, unii-m74yu7se79, rarechem aq a2 0002, pubchem3068, 3,5-dichlorobezaldehyde, 3,5-dichlorobenaldehyde, 3?5-dichlorobenzaldehyde, benzaldehyde,5-dichloro, acmc-1bxq6
PubChem CID 35746
IUPAC Name 3,5-dichlorobenzaldehyde
SMILES C1=C(C=C(C=C1Cl)Cl)C=O

Specifications

Melting Point 60°C to 66°C
Boiling Point 235°C to 240°C
Quantity 5 g
UN Number UN1759
Beilstein 2040553
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 175.01
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.