missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-pyridone-1-acetic acid, 98%, Thermo Scientific™

Product Code. 11447146
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447146 5 g 5g
11457146 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447146 Supplier Thermo Scientific Alfa Aesar Supplier No. B25471.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dichloro-4-pyridone-1-acetic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56187-37-2
Molecular Formula C7H5Cl2NO3
Molecular Weight (g/mol) 222.021
MDL Number MFCD02180547
InChI Key MFMXEEQESUEMSB-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-pyridone-1-acetic acid, 3,5-dichloro-4-pyridone-n-acetic acid, 2-3,5-dichloro-4-oxopyridin-1 4h-yl acetic acid, 1 4h-pyridineacetic acid, 3,5-dichloro-4-oxo, 3,5-dichloro-4-oxo-4h-pyridine-1-acetic acid, 3,5-dichloro-4-oxopyridin-1-yl acetic acid, 3,5-dichloro-4-pyridine-n-acetic acid, 2-3,5-dichloro-4-oxohydropyridyl acetic acid, pubchem11174, ksc272s7b
PubChem CID 92052
IUPAC Name 2-(3,5-dichloro-4-oxopyridin-1-yl)acetic acid
SMILES C1=C(C(=O)C(=CN1CC(=O)O)Cl)Cl

Specifications

Melting Point 239°C to 241°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 222.03
Percent Purity 98%
Chemical Name or Material 3,5-Dichloro-4-pyridone-1-acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.