missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-(hydroxymethyl)benzoic Acid, TRC

Product Code. 30385880
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385880 500 mg
30370657 1 g
30379213 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385880 Supplier Toronto Research Chemicals Supplier No. TRCD433790500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89894-53-1
Molecular Formula C8 H6 Cl2 O3
Molecular Weight (g/mol) 221.0374
Synonym p-Toluic acid, 3,5-dichloro-α-hydroxy- (7CI), 3,5-Dichloro-4-(hydroxymethyl)benzoic acid
IUPAC Name 3,5-dichloro-4-(hydroxymethyl)benzoic acid
SMILES OCc1c(Cl)cc(cc1Cl)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H6Cl2O3/c9-6-1-4(8(12)13)2-7(10)5(6)3-11/h1-2,11H,3H2,(H,12,13)
Formula Weight 219.9694
Chemical Name or Material 3,5-Dichloro-4-(hydroxymethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.