missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-hydroxybenzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number 11309006
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
13432-81-0
C6H3Cl3O3S
261.50
MFCD00171551
AELFLPJWVRGXKU-UHFFFAOYSA-N
2735994
OC1=C(Cl)C=C(C=C1Cl)S(Cl)(=O)=O
This item is not returnable. View return policy
13432-81-0
C6H3Cl3O3S
261.50
MFCD00171551
AELFLPJWVRGXKU-UHFFFAOYSA-N
2735994
OC1=C(Cl)C=C(C=C1Cl)S(Cl)(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed in the convergent syntheses involving hydroxyarenesulfonyl chlorides.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 118°C to 119°C
Quantity 5 g
UN Number UN3261
Beilstein 2806977
Sensitivity Moisture sensitive
Synonym 3,5-dichloro-4-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-4-hydroxybenzenesulphonyl chloride, 3,5-dichloro-4-hydroxy benzene sulphonyl chloride, 3,5-dichloro-4-hydroxy-benzenesulfonyl chloride, acmc-1c20j, 3,5-dichloro-4-hydroxyphenyl chlorosulfone, 3,5-dichloro-4-hydroxybenzenesulphonylchloride, benzenesulfonylchloride,3,5-dichloro-4-hydroxy, benzenesulfonyl chloride, 3,5-dichloro-4-hydroxy, benzenesulfonylchloride, 3,5-dichloro-4-hydroxy
Solubility Information Hydrolyzes in water.
IUPAC Name 3,5-dichloro-4-hydroxybenzenesulfonyl chloride
Formula Weight 261.51
Percent Purity 97%
Chemical Name or Material 3,5-Dichloro-4-hydroxybenzenesulfonyl chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'