missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-fluoro-1-nitrobenzene, 95%

Product Code. 15437848
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437848 25 g 25g
15427848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437848 Supplier Thermo Scientific Alfa Aesar Supplier No. H61321.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3107-19-5 | C6H2Cl2FNO2 | 209.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3107-19-5
Molecular Formula C6H2Cl2FNO2
Molecular Weight (g/mol) 209.99
MDL Number MFCD00129963
InChI Key VMAATSFMXSMKPG-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-fluoronitrobenzene, benzene, 1,3-dichloro-2-fluoro-5-nitro, 2,6-dichloro-4-nitrofluorobenzene, zlchem 371, pubchem4387, acmc-209hk6, ksc495g6p, 3,5-dichloro-4-fluoro-1-nitrobenzene, 1,3-dichloro-2-fluoro-5-nitro-benzene
PubChem CID 76554
IUPAC Name 1,3-dichloro-2-fluoro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Cl)=C(F)C(Cl)=C1

Specifications

Color White
Quantity 25 g
Formula Weight 209.99
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3,5-Dichloro-4-fluoro-1-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.