missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride, 90-95%, Thermo Scientific™

Product Code. 10721821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10721821 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10721821 Supplier Thermo Scientific Acros Supplier No. 163830250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 23378-88-3
Molecular Formula C6H3Cl3O3S
Molecular Weight (g/mol) 261.50
MDL Number MFCD00007432
InChI Key KXFQRJNVGBIDHA-UHFFFAOYSA-N
Synonym 3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone
PubChem CID 90078
SMILES OC1=C(Cl)C=C(Cl)C=C1S(Cl)(=O)=O

Specifikationer

CAS Min % 90.0
CAS Max % 100.0
Melting Point 78°C to 84°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 90% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula Cl2C6H2(OH)SO2Cl
Quantity 25 g
IUPAC Name 3,5-dichloro-2-hydroxybenzenesulfonyl chloride
Formula Weight 261.51
Percent Purity 90-95%
Physical Form Powder
Chemical Name or Material 3, 5-Dichloro-2-hydroxybenzenesulfonyl chloride, 90-95%
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 3,5-Dichloro-2-hydroxybenzenesulfonyl chloride
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faroangivelser
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.