missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dichloro-2,2,2-trifluoroacetophenone, TRC

Product Code. 30382909
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382909 25 g 25g
30384294 5 g 5g
30372802 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382909 Supplier Toronto Research Chemicals Supplier No. TRCD43640525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130336-16-2
Molecular Formula C8H3Cl2F3O
Molecular Weight (g/mol) 243.01
Synonym 1-(3,5-Dichlorophenyl)-2,2,2-trifluoroethanone, 1-(3',5'-Dichlorophenyl)-2,2,2-trifluoroethanone, 3',5'-Dichloro-2,2,2-trifluoroacetophenone, 1-(3,5-Dichlorophenyl)-2,2,2-trifluoro-ethanone
SMILES FC(F)(F)C(=O)c1cc(Cl)cc(Cl)c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H3Cl2F3O/c9-5-1-4(2-6(10)3-5)7(14)8(11,12)13/h1-3H
Formula Weight 241.9513
Chemical Name or Material 3',5'-Dichloro-2,2,2-trifluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.