missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromosalicylic acid, 98%, Thermo Scientific™

Product Code. 11491657
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491657 25 g 25g
11401667 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491657 Supplier Thermo Scientific Alfa Aesar Supplier No. B25642.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3147-55-5
Molecular Formula C7H4Br2O3
Molecular Weight (g/mol) 295.914
MDL Number MFCD00002441
InChI Key BFBZHSOXKROMBG-UHFFFAOYSA-N
Synonym 3,5-dibromosalicylic acid, benzoic acid, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzoic acid, unii-t5jo5ua21h, salicylic acid, 3,5-dibromo, t5jo5ua21h, rarechem al bo 2002, attercop-chm at113669, labotest-bb lt00455167, labotest-bb lt03333482
PubChem CID 18464
IUPAC Name 3,5-dibromo-2-hydroxybenzoic acid
SMILES C1=C(C=C(C(=C1Br)O)C(=O)O)Br

Specifications

Melting Point 223°C to 228°C
Quantity 25 g
Beilstein 2210805
Merck Index 14,3028
Formula Weight 295.92
Percent Purity 98%
Chemical Name or Material 3,5-Dibromosalicylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.