missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromosalicylaldehyde, 98%

Product Code. 11474744
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474744 10 g 10g
11484744 50 g 50g
11494744 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474744 Supplier Thermo Scientific Alfa Aesar Supplier No. A13729.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-59-5 | C7H4Br2O2 | 279.915 g/mol

3,5-Dibromosalicylaldehyde reacts with alkyl cyanoacetates in the presence of ammonium acetate to yield 4H- chromenes. It can be used in the synthesis of Schiff base and can be used as reactant for synthesis of Schiff base ligands which forms mononuclear complexes with copper(II), nickel(II), zinc(II) and cobalt(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromosalicylaldehyde reacts with alkyl cyanoacetates in the presence of ammonium acetate to yield 4H- chromenes. It can be used in the synthesis of Schiff base and can be used as reactant for synthesis of Schiff base ligands which forms mononuclear complexes with copper(II), nickel(II), zinc(II) and cobalt(II).

Solubility
Soluble in methanol 25 mg/mL. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-59-5
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.915
MDL Number MFCD00003318
InChI Key JHZOXYGFQMROFJ-UHFFFAOYSA-N
Synonym 3,5-dibromosalicylaldehyde, dalyde, 3,5-dibromosalicylaldeyde, benzaldehyde, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzaldehyde, 3,5-dibromosilicylaldehyde, unii-7a260pbi3a, salicylaldehyde, 3,5-dibromo, 3,5-dibromsalicylaldehyd, 3,5-dibromo salicyclic aldehyde
PubChem CID 7024
IUPAC Name 3,5-dibromo-2-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1Br)O)C=O)Br

Specifications

Melting Point 81°C to 84°C
Quantity 10 g
Beilstein 1424739
Sensitivity Air Sensitive
Merck Index 14,3027
Solubility Information Soluble in methanol 25mg/mL. Insoluble in water.
Formula Weight 279.93
Percent Purity 98%
Chemical Name or Material 3,5-Dibromosalicylaldehyde
compliance-icons
Product Identifier
  • 3,5-Dibromosalicylaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3,5-Dibromo-2-hydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.